accessibility menu, dialog, popup

UserName

H3COFO

CAS RN 105728-90-3

IUPAC Name: 2-fluoro-5-methoxybenzaldehyde
Synonyms: 2-fluoro-5-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: DKIQXHIAEMGZGO-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(C=O)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)