missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2-naphthaleneboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 352525-98-5
Molecular Formula C12H13BO3
Molecular Weight (g/mol) 216.04
InChI Key INXXVGFSXYJGHI-UHFFFAOYSA-N
Synonym 6-ethoxy-2-naphthaleneboronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, boronic acid, 6-ethoxy-2-naphthalenyl, 2-ethoxy-6-naphthaleneboronic acid, boronic acid, b-6-ethoxy-2-naphthalenyl, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxy-2, acmc-209ies, boronic acid,b-6-ethoxy-2-naphthalenyl
PubChem CID 4641693
IUPAC Name (6-ethoxynaphthalen-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C=C(C=C2)OCC)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC440230010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

352525-98-5
216.04
6-ethoxy-2-naphthaleneboronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, boronic acid, 6-ethoxy-2-naphthalenyl, 2-ethoxy-6-naphthaleneboronic acid, boronic acid, b-6-ethoxy-2-naphthalenyl, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxy-2, acmc-209ies, boronic acid,b-6-ethoxy-2-naphthalenyl
(6-ethoxynaphthalen-2-yl)boronic acid
C12H13BO3
INXXVGFSXYJGHI-UHFFFAOYSA-N
4641693
B(C1=CC2=C(C=C1)C=C(C=C2)OCC)(O)O

Specifications

352525-98-5
100.0
96% min. (HPLC)
C12H13BO3
INXXVGFSXYJGHI-UHFFFAOYSA-N
(6-ethoxynaphthalen-2-yl)boronic acid
4641693
97%
96.0
Authentic
Glass Bottle
6-ethoxy-2-naphthaleneboronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, boronic acid, 6-ethoxy-2-naphthalenyl, 2-ethoxy-6-naphthaleneboronic acid, boronic acid, b-6-ethoxy-2-naphthalenyl, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxy-2, acmc-209ies, boronic acid,b-6-ethoxy-2-naphthalenyl
B(C1=CC2=C(C=C1)C=C(C=C2)OCC)(O)O
216.04
216.04
6-Ethoxy-2-naphthaleneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only