accessibility menu, dialog, popup

UserName

CAS RN 352525-98-5

H3COBOHOH

CAS RN 352525-98-5

IUPAC Name: (6-ethoxynaphthalen-2-yl)boronic acid
Synonyms: 6-ethoxynaphthalen-2-ylboronic acid
Molecular Weight (g/mol): 216.04
Molecular Formula: C12H13BO3
InChi Key: INXXVGFSXYJGHI-UHFFFAOYSA-N
SMILES: CCOC1=CC2=CC=C(C=C2C=C1)B(O)O
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results