missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Dimethylamino-1-hexanol 97.0+%, TCI America™
Supplier: TCI America D166425ML
Specifications
| 6-Dimethylamino-1-hexanol | |
| 117°C | |
| MFCD00046028 | |
| 2735 | |
| CN(C)CCCCCCO | |
| 145.25 | |
| 145.25 | |
| Reagent |
| 1862-07-3 | |
| C8H19NO | |
| 25 mL | |
| QCXNXRUTKSIZND-UHFFFAOYSA-N | |
| 6-(dimethylamino)hexan-1-ol | |
| 74623 | |
| ≥97.0% (GC) | |
| Liquid |
Chemical Identifiers
| 1862-07-3 | |
| 145.25 | |
| QCXNXRUTKSIZND-UHFFFAOYSA-N | |
| 6-(dimethylamino)hexan-1-ol |
| C8H19NO | |
| MFCD00046028 | |
| 74623 | |
| CN(C)CCCCCCO |
Safety and Handling
EINECSNumber : (2)-3530
TSCA : Yes