missing translation for 'onlineSavingsMsg'
Learn More

6-Dimethylamino-1-hexanol 97.0+%, TCI America™

Catalog No. p-7126861
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16645ML 5 mL
D166425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16645ML Supplier TCI America Supplier No. D16645ML

Chemical Identifiers

CAS 1862-07-3
Molecular Formula C8H19NO
Molecular Weight (g/mol) 145.25
MDL Number MFCD00046028
InChI Key QCXNXRUTKSIZND-UHFFFAOYSA-N
PubChem CID 74623
IUPAC Name 6-(dimethylamino)hexan-1-ol
SMILES CN(C)CCCCCCO

Specifications

Boiling Point 117°C
Quantity 5 mL
UN Number 2735
Formula Weight 145.25
Percent Purity ≥97.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 6-Dimethylamino-1-hexanol
EINECSNumber (2)-3530
TSCA Yes