accessibility menu, dialog, popup

UserName

CAS RN 1862-07-3

H3CNCH3OH

CAS RN 1862-07-3

IUPAC Name: 6-(dimethylamino)hexan-1-ol
Synonyms: 6-(dimethylamino)hexan-1-ol
Molecular Weight (g/mol): 145.25
Molecular Formula: C8H19NO
InChi Key: QCXNXRUTKSIZND-UHFFFAOYSA-N
SMILES: CN(C)CCCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results