missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-chloronicotinamide, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 6271-78-9 |
|---|---|
| Molecular Formula | C6H5ClN2O |
| Molecular Weight (g/mol) | 156.57 |
| MDL Number | MFCD00006242 |
| InChI Key | ZIJAZUBWHAZHPL-UHFFFAOYSA-N |
| Synonym | 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t |
| PubChem CID | 80456 |
| IUPAC Name | 6-chloropyridine-3-carboxamide |
| SMILES | NC(=O)C1=CC=C(Cl)N=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC109560010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H5ClN2O | |
| MFCD00006242 | |
| 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t | |
| 6-chloropyridine-3-carboxamide |
Specifications
| 6271-78-9 | |
| 100.0 | |
| 98% | |
| C6H5ClN2O | |
| MFCD00006242 | |
| 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t | |
| NC(=O)C1=CC=C(Cl)N=C1 | |
| 156.57 | |
| 156.57 | |
| 6-Chloronicotinamide |
| 97.5 | |
| Authentic | |
| Glass Bottle | |
| Cl(C5H3N)CONH2 | |
| 22,44 | |
| ZIJAZUBWHAZHPL-UHFFFAOYSA-N | |
| 6-chloropyridine-3-carboxamide | |
| 80456 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 228-456-8
RUO – Research Use Only