accessibility menu, dialog, popup

UserName

H2NOClN

CAS RN 6271-78-9

IUPAC Name: 6-chloropyridine-3-carboxamide
Synonyms: 6-chloropyridine-3-carboxamide
Molecular Weight (g/mol): 156.57
Molecular Formula: C6H5ClN2O
InChi Key: ZIJAZUBWHAZHPL-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(Cl)N=C1
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)