missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-3(2H)-pyridazinone 98.0+%, TCI America™
Supplier: TCI America C237725G
Specifications
| 6-Chloro-3(2H)-pyridazinone | |
| 142°C | |
| C4H3ClN2O | |
| 25 g | |
| YICPBKWYZXFJNB-UHFFFAOYSA-N | |
| 3-chloro-1H-pyridazin-6-one | |
| 252828 | |
| ≥98.0% (GC,T) |
| 19064-67-6 | |
| White-Yellow | |
| MFCD00233955 | |
| 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol | |
| C1=CC(=NNC1=O)Cl | |
| 130.531 | |
| 130.53 | |
| Crystalline Powder |
Chemical Identifiers
| 19064-67-6 | |
| 130.531 | |
| YICPBKWYZXFJNB-UHFFFAOYSA-N | |
| 252828 | |
| C1=CC(=NNC1=O)Cl |
| C4H3ClN2O | |
| MFCD00233955 | |
| 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol | |
| 3-chloro-1H-pyridazin-6-one |
Safety and Handling
TSCA : No