missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. p-7125438
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23775G 5 g
C237725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23775G Supplier TCI America Supplier No. C23775G
Only null left

Chemical Identifiers

CAS 19064-67-6
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD00233955
InChI Key YICPBKWYZXFJNB-UHFFFAOYSA-N
Synonym 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
PubChem CID 252828
IUPAC Name 3-chloro-1H-pyridazin-6-one
SMILES C1=CC(=NNC1=O)Cl

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 130.53
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3(2H)-pyridazinone