accessibility menu, dialog, popup

UserName

ClNNHO

CAS RN 19064-67-6

IUPAC Name: 6-chloro-2,3-dihydropyridazin-3-one
Synonyms: 6-chloro-2H-pyridazin-3-one
Molecular Weight (g/mol): 130.53
Molecular Formula: C4H3ClN2O
InChi Key: YICPBKWYZXFJNB-UHFFFAOYSA-N
SMILES: ClC1=NNC(=O)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)