missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 944896-42-8 |
|---|---|
| Molecular Formula | C8H5BrN2O2 |
| Molecular Weight (g/mol) | 241.04 |
| MDL Number | MFCD10698063 |
| InChI Key | SDKJLYHPCBMDAD-UHFFFAOYSA-N |
| Synonym | 6-bromoimidazo 1,2-a pyridine-3-carboxylic acid, 6-bromoimidazo 1,2-a pyridine-3-carboxylicacid, 6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-3-carboxyimidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo |
| PubChem CID | 22031912 |
| IUPAC Name | 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid |
| SMILES | OC(=O)C1=CN=C2C=CC(Br)=CN12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC449910050
|
N/A | 5g |
N/A
|
N/A | |||||
Chemical Identifiers
| 944896-42-8 | |
| 241.04 | |
| SDKJLYHPCBMDAD-UHFFFAOYSA-N | |
| 22031912 | |
| OC(=O)C1=CN=C2C=CC(Br)=CN12 |
| C8H5BrN2O2 | |
| MFCD10698063 | |
| 6-bromoimidazo 1,2-a pyridine-3-carboxylic acid, 6-bromoimidazo 1,2-a pyridine-3-carboxylicacid, 6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-3-carboxyimidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo | |
| 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid |
Specifications
| 944896-42-8 | |
| 96% min. (HPLC) | |
| MFCD10698063 | |
| SDKJLYHPCBMDAD-UHFFFAOYSA-N | |
| 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid | |
| 22031912 | |
| 97% |
| Authentic | |
| C8H5BrN2O2 | |
| 6-bromoimidazo 1,2-a pyridine-3-carboxylic acid, 6-bromoimidazo 1,2-a pyridine-3-carboxylicacid, 6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-3-carboxyimidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo | |
| OC(=O)C1=CN=C2C=CC(Br)=CN12 | |
| 241.04 | |
| 241.04 | |
| 6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only