missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 5g |
| Formula Weight | 241.04 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. |
RUO – Research Use Only