missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromochromone 98.0+%, TCI America™
$160.30 - $503.30
Chemical Identifiers
| CAS | 51483-92-2 |
|---|---|
| Molecular Formula | C9H5BrO2 |
| Molecular Weight (g/mol) | 225.041 |
| MDL Number | MFCD00239369 |
| InChI Key | XVNBWGGBXOJIDR-UHFFFAOYSA-N |
| Synonym | 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran |
| PubChem CID | 548522 |
| IUPAC Name | 6-bromochromen-4-one |
| SMILES | C1=CC2=C(C=C1Br)C(=O)C=CO2 |
Chemical Identifiers
| 51483-92-2 | |
| 225.041 | |
| XVNBWGGBXOJIDR-UHFFFAOYSA-N | |
| 548522 | |
| C1=CC2=C(C=C1Br)C(=O)C=CO2 |
| C9H5BrO2 | |
| MFCD00239369 | |
| 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran | |
| 6-bromochromen-4-one |
Specifications
| 51483-92-2 | |
| White-Yellow | |
| MFCD00239369 | |
| 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran | |
| C1=CC2=C(C=C1Br)C(=O)C=CO2 | |
| 225.041 | |
| 225.04 | |
| Crystalline Powder |
| 138°C | |
| C9H5BrO2 | |
| 5 g | |
| XVNBWGGBXOJIDR-UHFFFAOYSA-N | |
| 6-bromochromen-4-one | |
| 548522 | |
| ≥98.0% (GC) | |
| 6-Bromochromone |
Safety and Handling
TSCA : No