CAS RN 51483-92-2
CAS RN 51483-92-2
IUPAC Name:
6-bromo-4H-chromen-4-one
Synonyms:
6-bromochromen-4-one
Molecular Weight (g/mol):
225.04
Molecular Formula:
C9H5BrO2
InChi Key:
XVNBWGGBXOJIDR-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2OC=CC(=O)C2=C1
1
–
1
of
1
results
1
6-Bromochromone 98.0+%, TCI America™
CAS: 51483-92-2 Molecular Formula: C9H5BrO2 Molecular Weight (g/mol): 225.041 MDL Number: MFCD00239369 InChI Key: XVNBWGGBXOJIDR-UHFFFAOYSA-N Synonym: 6-bromochromone,6-bromo-4h-chromen-4-one,4h-1-benzopyran-4-one,6-bromo,4h-1-benzopyran-4-one, 6-bromo,acmc-1aser,6-bromanylchromen-4-one,6-bromo-1-benzopyran-4-one,6-bromo-4h-chromen-4-one #,6-bromo-4-oxo-4h-1-benzopyran PubChem CID: 548522 IUPAC Name: 6-bromochromen-4-one SMILES: C1=CC2=C(C=C1Br)C(=O)C=CO2