missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromochromone 98.0+%, TCI America™
Supplier: TCI America B333525G
Specifications
| 6-Bromochromone | |
| 138°C | |
| C9H5BrO2 | |
| 25 g | |
| XVNBWGGBXOJIDR-UHFFFAOYSA-N | |
| 6-bromochromen-4-one | |
| 548522 | |
| ≥98.0% (GC) |
| 51483-92-2 | |
| White-Yellow | |
| MFCD00239369 | |
| 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran | |
| C1=CC2=C(C=C1Br)C(=O)C=CO2 | |
| 225.041 | |
| 225.04 | |
| Crystalline Powder |
Chemical Identifiers
| 51483-92-2 | |
| 225.041 | |
| XVNBWGGBXOJIDR-UHFFFAOYSA-N | |
| 548522 | |
| C1=CC2=C(C=C1Br)C(=O)C=CO2 |
| C9H5BrO2 | |
| MFCD00239369 | |
| 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran | |
| 6-bromochromen-4-one |
Safety and Handling
TSCA : No