missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-2-naphthoic Acid 98.0+%, TCI America™
Supplier: TCI America B38235G
Specifications
| 6-Bromo-2-naphthoic Acid | |
| 297°C | |
| C11H7BrO2 | |
| 5 g | |
| NPMCAVBMOTZUPD-UHFFFAOYSA-N | |
| 6-bromonaphthalene-2-carboxylic acid | |
| 4549852 | |
| ≥98.0% (GC,T) |
| 5773-80-8 | |
| White-Yellow | |
| MFCD01075720 | |
| 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid | |
| C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O | |
| 251.079 | |
| 251.08 | |
| Crystalline Powder |
Chemical Identifiers
| 5773-80-8 | |
| 251.079 | |
| NPMCAVBMOTZUPD-UHFFFAOYSA-N | |
| 4549852 | |
| C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O |
| C11H7BrO2 | |
| MFCD01075720 | |
| 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid | |
| 6-bromonaphthalene-2-carboxylic acid |
Safety and Handling
TSCA : No