missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthoic Acid 98.0+%, TCI America™

Catalog No. p-7123184
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38235G 5 g
B382325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38235G Supplier TCI America Supplier No. B38235G
Only null left

Chemical Identifiers

CAS 5773-80-8
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD01075720
InChI Key NPMCAVBMOTZUPD-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid
PubChem CID 4549852
IUPAC Name 6-bromonaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O

Specifications

Melting Point 297°C
Color White-Yellow
Quantity 5 g
Formula Weight 251.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-naphthoic Acid