Learn More
6-Bromo-2-naphthoic acid, 98%
CAS: 5773-80-8 | C11H7BrO2 | 251.079 g/mol
Supplier: Thermo Scientific Chemicals L1609314
Description
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 6-Bromo-2-naphthoic acid | |
| 292°C to 296°C | |
| MFCD01075720 | |
| 2328940 | |
| Insoluble in water. | |
| C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O | |
| 251.079 | |
| 251.08 |
| 5773-80-8 | |
| C11H7BrO2 | |
| 25 g | |
| 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid | |
| NPMCAVBMOTZUPD-UHFFFAOYSA-N | |
| 6-bromonaphthalene-2-carboxylic acid | |
| 4549852 | |
| 98% |
Chemical Identifiers
| 5773-80-8 | |
| 251.079 | |
| NPMCAVBMOTZUPD-UHFFFAOYSA-N | |
| 4549852 | |
| C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O |
| C11H7BrO2 | |
| MFCD01075720 | |
| 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid | |
| 6-bromonaphthalene-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only