missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthoic acid, 98%

Catalog No. p-7046734
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1609306 5 g
AAL1609314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1609306 Supplier Thermo Scientific Chemicals Supplier No. L1609306

CAS: 5773-80-8 | C11H7BrO2 | 251.079 g/mol

Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 5773-80-8
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD01075720
InChI Key NPMCAVBMOTZUPD-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid
PubChem CID 4549852
IUPAC Name 6-bromonaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O

Specifications

Melting Point 292°C to 296°C
Quantity 5 g
Beilstein 2328940
Solubility Information Insoluble in water.
Formula Weight 251.08
Percent Purity 98%
Chemical Name or Material 6-Bromo-2-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only