missing translation for 'onlineSavingsMsg'
Learn More

6-Benzylamino-1-hexanol 97.0+%, TCI America™

Catalog No. B29685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29685G 5 g
B296825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29685G Supplier TCI America Supplier No. B29685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133437-08-8
Molecular Formula C13H21NO
Molecular Weight (g/mol) 207.32
MDL Number MFCD09038506
InChI Key ILKSKQBXMQDBFT-UHFFFAOYSA-N
Synonym N-(6-Hydroxyhexyl)benzylamine
PubChem CID 15133217
IUPAC Name 6-(benzylamino)hexan-1-ol
SMILES OCCCCCCNCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 207.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Benzylamino-1-hexanol