accessibility menu, dialog, popup

UserName

NHOH

CAS RN 133437-08-8

IUPAC Name: 6-(benzylamino)hexan-1-ol
Synonyms: 6-(benzylamino)hexan-1-ol
Molecular Weight (g/mol): 207.32
Molecular Formula: C13H21NO
InChi Key: ILKSKQBXMQDBFT-UHFFFAOYSA-N
SMILES: OCCCCCCNCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results