missing translation for 'onlineSavingsMsg'
Learn More

6-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-norleucine 98.0+%, TCI America™

Catalog No. A3023250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3023250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3023250MG Supplier TCI America Supplier No. A3023250MG
Only null left

Chemical Identifiers

CAS 159610-89-6
Molecular Formula C21H22N4O4
Molecular Weight (g/mol) 394.431
MDL Number MFCD13182319
InChI Key PJRFTUILPGJJIO-IBGZPJMESA-N
Synonym 6-Azido-N-Fmoc-L-norleucine
PubChem CID 25146063
IUPAC Name (2S)-6-azido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=[N+]=[N-])C(=O)O

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 250 mg
Formula Weight 394.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-norleucine
TSCA No
Recommended Storage Refrigerator