accessibility menu, dialog, popup

UserName

OONHNNNOHO

CAS RN 159610-89-6

IUPAC Name: 6-azido-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-azido-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 394.43
Molecular Formula: C21H22N4O4
InChi Key: PJRFTUILPGJJIO-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCN=[N+]=[N-])NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results