missing translation for 'onlineSavingsMsg'
Learn More

6-Azauridine

CAS: 54-25-1 | C8H11N3O6 | 245.191 g/mol

Supplier:  Thermo Scientific Chemicals J6130906

 View more versions of this product

Catalog No. AAJ6130906


Only null left

Description

Description

6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Specifications

Specifications

6-Azauridine
160°C to 161°C
C8H11N3O6
5 g
6-azauridine, 6-azuridine, azauridine, azur, riboazauracil, riboazauratsil, 6-azauracil 1-riboside, ribo-azuracil, 6-azauracil riboside, 6-azur
WYXSYVWAUAUWLD-SHUUEZRQSA-N
2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazine-3,5-dione
5901
245.19
54-25-1
White
MFCD00006472
14,904
It is soluble in water.
C1=NN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
245.191
CHEBI:35668
Powder

Chemical Identifiers

54-25-1
245.191
WYXSYVWAUAUWLD-SHUUEZRQSA-N
5901
2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazine-3,5-dione
C8H11N3O6
MFCD00006472
6-azauridine, 6-azuridine, azauridine, azur, riboazauracil, riboazauratsil, 6-azauracil 1-riboside, ribo-azuracil, 6-azauracil riboside, 6-azur
CHEBI:35668
C1=NN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H312-H332
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H351

EINECSNumber : 200-199-6

RTECSNumber : XY8575000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only