missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinamide 99.0+%, TCI America™

Supplier:  TCI America A09865G

 View more versions of this product

Catalog No. A09865G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

6-Aminonicotinamide
White-Yellow
MFCD00006327
2811
ZLWYEPMDOUQDBW-UHFFFAOYSA-N
6-aminopyridine-3-carboxamide
9500
137.14
Crystalline Powder
329-89-5
C6H7N3O
5 g
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
NC(=O)C1=CC=C(N)N=C1
137.14
CHEBI:74514
≥99.0% (T)

Chemical Identifiers

329-89-5
137.14
ZLWYEPMDOUQDBW-UHFFFAOYSA-N
9500
6-aminopyridine-3-carboxamide
C6H7N3O
MFCD00006327
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
CHEBI:74514
NC(=O)C1=CC=C(N)N=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : US4550000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions