accessibility menu, dialog, popup

UserName

CAS RN 329-89-5

IUPAC Name: 6-aminopyridine-3-carboxamide
Synonyms: 6-aminonicotinamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: ZLWYEPMDOUQDBW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(N)N=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)