missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinamide 99.0+%, TCI America™

Catalog No. p-7117169
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09861G 1 g
A09865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09861G Supplier TCI America Supplier No. A09861G
Only null left

Chemical Identifiers

CAS 329-89-5
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006327
InChI Key ZLWYEPMDOUQDBW-UHFFFAOYSA-N
Synonym 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
PubChem CID 9500
ChEBI CHEBI:74514
IUPAC Name 6-aminopyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(N)N=C1

Specifications

Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 137.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminonicotinamide
RTECSNumber US4550000
TSCA No