missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic acid, 99%

6-aminohexanoic acid, CAS # 60-32-2, is a synthetic lysine derivative with inhibitory activity against fibrinolysis. It shows hematologic activity. | CAS: 60-32-2 | C6H13NO2 | 131.175 g/mol

$368.88 - $1,442.27

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00008238
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1471936
View Documents Promotion Details
N/A 500 g
Each for $368.88
Only null left
 
AAA147190E
View Documents Promotion Details
N/A 2500 g
Each for $1,442.27
Only null left
 
Description

Description

6-Aminohexanoic acid is used as an intermediate in the polymerization of Nylon-6. It is involved in the extraction of aldehydes from the reaction mixture as well as to improve the solubilization of membrane proteins in electrophoresis. It acts as an inhibitor of fibrinolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 6-aminohexanoic acid, an epsilon-amino acid, shows an antifibrinolytic and hematologic activity
  • This compound competitively inhibits the activation of plasminogen; it consequently reduces the conversion of plasminogen to plasmin (fibrinolysin), an enzyme responsible for the degradation of fibrin clots, fibrinogen, and other plasma proteins, including the procoagulant factors V and VIII

Application

  • It has been used as an intermediate in the polymerization of Nylon-6
  • It is used for biomolecular applications to extract aldehydes from the reaction mixture and to increase membrane protein solubilization in electrophoresis experiments
  • It has been described to inhibit plasminogen binding to activated platelets
  • This compound can bind and inactivate carboxypeptidase B
  • This compound can also inhibit the activation of the first component of the complement system
Specifications

Chemical Identifiers

60-32-2
131.175
SLXKOJJOQWFEFD-UHFFFAOYSA-N
564
6-aminohexanoic acid
C6H13NO2
MFCD00008238
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
CHEBI:16586
C(CCC(=O)O)CCN

Specifications

60-32-2
≥99%
H2N(CH2)5CO2H
906872
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminohexanoic acid
564
131.18
6-Aminohexanoic acid
210°C to 212°C
C6H13NO2
MFCD00008238
14,432
Soluble in water. Slightly soluble in alcohol. Practically insoluble in ether and chloroform.
C(CCC(=O)O)CCN
131.175
CHEBI:16586
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-469-3

RTECSNumber : MO6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only