missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic acid, 99%

6-aminohexanoic acid, CAS # 60-32-2, is a synthetic lysine derivative with inhibitory activity against fibrinolysis. It shows hematologic activity. | CAS: 60-32-2 | C6H13NO2 | 131.175 g/mol

Supplier:  Thermo Scientific Chemicals A147190E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA147190E


Only null left

Alert:

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.


Description

Description

6-Aminohexanoic acid is used as an intermediate in the polymerization of Nylon-6. It is involved in the extraction of aldehydes from the reaction mixture as well as to improve the solubilization of membrane proteins in electrophoresis. It acts as an inhibitor of fibrinolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 6-aminohexanoic acid, an epsilon-amino acid, shows an antifibrinolytic and hematologic activity
  • This compound competitively inhibits the activation of plasminogen; it consequently reduces the conversion of plasminogen to plasmin (fibrinolysin), an enzyme responsible for the degradation of fibrin clots, fibrinogen, and other plasma proteins, including the procoagulant factors V and VIII

Application

  • It has been used as an intermediate in the polymerization of Nylon-6
  • It is used for biomolecular applications to extract aldehydes from the reaction mixture and to increase membrane protein solubilization in electrophoresis experiments
  • It has been described to inhibit plasminogen binding to activated platelets
  • This compound can bind and inactivate carboxypeptidase B
  • This compound can also inhibit the activation of the first component of the complement system
Specifications

Chemical Identifiers

60-32-2
131.175
SLXKOJJOQWFEFD-UHFFFAOYSA-N
564
6-aminohexanoic acid
C6H13NO2
MFCD00008238
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
CHEBI:16586
C(CCC(=O)O)CCN

Specifications

6-Aminohexanoic acid
210°C to 212°C
C6H13NO2
2500 g
906872
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminohexanoic acid
564
131.18
60-32-2
≥99%
H2N(CH2)5CO2H
MFCD00008238
14,432
Soluble in water. Slightly soluble in alcohol. Practically insoluble in ether and chloroform.
C(CCC(=O)O)CCN
131.175
CHEBI:16586
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-469-3

RTECSNumber : MO6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions