missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-5-bromoquinoxaline 98.0+%, TCI America™
$219.95 - $874.24
Chemical Identifiers
| CAS | 50358-63-9 |
|---|---|
| Molecular Formula | C8H6BrN3 |
| Molecular Weight (g/mol) | 224.06 |
| MDL Number | MFCD00799024 |
| InChI Key | IRAOSCSPAYZRJE-UHFFFAOYSA-N |
| PubChem CID | 14928442 |
| IUPAC Name | 5-bromoquinoxalin-6-amine |
| SMILES | NC1=CC=C2N=CC=NC2=C1Br |
Chemical Identifiers
| 50358-63-9 | |
| 224.06 | |
| IRAOSCSPAYZRJE-UHFFFAOYSA-N | |
| 5-bromoquinoxalin-6-amine |
| C8H6BrN3 | |
| MFCD00799024 | |
| 14928442 | |
| NC1=CC=C2N=CC=NC2=C1Br |
Specifications
| 50358-63-9 | |
| C8H6BrN3 | |
| 5 g | |
| NC1=CC=C2N=CC=NC2=C1Br | |
| 224.06 | |
| 224.06 | |
| Crystalline Powder |
| 157°C | |
| MFCD00799024 | |
| IRAOSCSPAYZRJE-UHFFFAOYSA-N | |
| 5-bromoquinoxalin-6-amine | |
| 14928442 | |
| ≥98.0% (GC,T) | |
| 6-Amino-5-bromoquinoxaline |
Safety and Handling
TSCA : No