accessibility menu, dialog, popup

UserName

NNH2NBr

CAS RN 50358-63-9

IUPAC Name: 5-bromoquinoxalin-6-amine
Synonyms: 5-bromoquinoxalin-6-amine
Molecular Weight (g/mol): 224.06
Molecular Formula: C8H6BrN3
InChi Key: IRAOSCSPAYZRJE-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=CC=NC2=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results