missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-bromoquinoxaline 98.0+%, TCI America™

Catalog No. A29135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29135G 5 g
A291325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29135G Supplier TCI America Supplier No. A29135G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50358-63-9
Molecular Formula C8H6BrN3
Molecular Weight (g/mol) 224.06
MDL Number MFCD00799024
InChI Key IRAOSCSPAYZRJE-UHFFFAOYSA-N
PubChem CID 14928442
IUPAC Name 5-bromoquinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1Br

Specifications

Melting Point 157°C
Quantity 5 g
Formula Weight 224.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-5-bromoquinoxaline