missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-naphthol-3-sulfonic Acid Hydrate 95.0+%, TCI America™

Catalog No. p-7116860
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A037025G 25 g
A0370500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A037025G Supplier TCI America Supplier No. A037025G

Chemical Identifiers

CAS 87-02-5
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00003970
InChI Key KYARBIJYVGJZLB-UHFFFAOYSA-N
Synonym 7-Amino-4-hydroxy-2-naphthalenesulfonic Acid, J Acid
PubChem CID 6868
ChEBI CHEBI:87316
IUPAC Name 7-amino-4-hydroxynaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C(C=C2C=C1N)S(=O)(=O)O)O

Specifications

Quantity 25 g
Formula Weight 239.25
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-naphthol-3-sulfonic Acid Hydrate
EINECSNumber (4)-0514
RTECSNumber QK1295500
TSCA Yes