missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-naphthalenesulfonic Acid 97.0+%, TCI America™

Catalog No. A034725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A034725G 25 g
A0347500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A034725G Supplier TCI America Supplier No. A034725G

Chemical Identifiers

CAS 81-05-0
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00035724
InChI Key YUNBHHWDQDGWHC-UHFFFAOYSA-N
PubChem CID 66470
IUPAC Name 6-aminonaphthalene-1-sulfonic acid
SMILES NC1=CC=C2C(C=CC=C2S(O)(=O)=O)=C1

Specifications

Color Red-White
Quantity 25 g
Synonym Dahl′Cs Acid, 2-Naphthylamine-5-sulfonic Acid
Formula Weight 223.25
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-naphthalenesulfonic Acid
EINECSNumber (4)-0493
TSCA No