accessibility menu, dialog, popup

UserName

CAS RN 81-05-0

IUPAC Name: 6-aminonaphthalene-1-sulfonic acid
Synonyms: 6-aminonaphthalene-1-sulfonic acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: YUNBHHWDQDGWHC-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(C=CC=C2S(O)(=O)=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results