missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-2-benzoxazolinone 98.0+%, TCI America™

Catalog No. A29071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29071G 1 g
A29075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29071G Supplier TCI America Supplier No. A29071G

Chemical Identifiers

CAS 54903-09-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD01664312
InChI Key QXBNAXVXLAHDTE-UHFFFAOYSA-N
Synonym 6-acetyl-2 3h-benzoxazolone, 2 3h-benzoxazolone, 6-acetyl, jpb 2, 6-acetyl-2-benzoxazolinone, 6-acetylbenzo d oxazol-2 3h-one, 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one, acetyl-6 benzoxazolinone french, 6-acetyl-3h-benzooxazol-2-one, 6-acetyl-1,3-benzoxazol-2 3h-one, 6-acetyl-2-oxo-3-hydrobenzoxazole
PubChem CID 689054
IUPAC Name 6-acetyl-3H-1,3-benzoxazol-2-one
SMILES CC(=O)C1=CC2=C(C=C1)NC(=O)O2

Specifications

Melting Point 236°C
Color White
Quantity 1 g
Formula Weight 177.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetyl-2-benzoxazolinone
RTECSNumber DM5285500
TSCA No