accessibility menu, dialog, popup

UserName

CAS RN 54903-09-2

IUPAC Name: 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-acetyl-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: QXBNAXVXLAHDTE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2NC(=O)OC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)