missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Acetyl-2(3H)-benzoxazolone, 97%, Thermo Scientific Chemicals
$294.81 - $294.81
Chemical Identifiers
| CAS | 54903-09-2 |
|---|---|
| Molecular Formula | C9H7NO3 |
| Molecular Weight (g/mol) | 177.159 |
| MDL Number | MFCD01664312 |
| InChI Key | QXBNAXVXLAHDTE-UHFFFAOYSA-N |
| Synonym | 6-acetyl-2 3h-benzoxazolone, 2 3h-benzoxazolone, 6-acetyl, jpb 2, 6-acetyl-2-benzoxazolinone, 6-acetylbenzo d oxazol-2 3h-one, 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one, acetyl-6 benzoxazolinone french, 6-acetyl-3h-benzooxazol-2-one, 6-acetyl-1,3-benzoxazol-2 3h-one, 6-acetyl-2-oxo-3-hydrobenzoxazole |
| PubChem CID | 689054 |
| IUPAC Name | 6-acetyl-3H-1,3-benzoxazol-2-one |
| SMILES | CC(=O)C1=CC2=C(C=C1)NC(=O)O2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2736806
|
Thermo Scientific Chemicals
H2736806 |
5 g |
Each for $294.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54903-09-2 | |
| 177.159 | |
| QXBNAXVXLAHDTE-UHFFFAOYSA-N | |
| 689054 | |
| CC(=O)C1=CC2=C(C=C1)NC(=O)O2 |
| C9H7NO3 | |
| MFCD01664312 | |
| 6-acetyl-2 3h-benzoxazolone, 2 3h-benzoxazolone, 6-acetyl, jpb 2, 6-acetyl-2-benzoxazolinone, 6-acetylbenzo d oxazol-2 3h-one, 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one, acetyl-6 benzoxazolinone french, 6-acetyl-3h-benzooxazol-2-one, 6-acetyl-1,3-benzoxazol-2 3h-one, 6-acetyl-2-oxo-3-hydrobenzoxazole | |
| 6-acetyl-3H-1,3-benzoxazol-2-one |
Specifications
| 54903-09-2 | |
| C9H7NO3 | |
| 5 g | |
| QXBNAXVXLAHDTE-UHFFFAOYSA-N | |
| 6-acetyl-3H-1,3-benzoxazol-2-one | |
| 689054 | |
| 97% |
| 232°C to 239°C | |
| MFCD01664312 | |
| 6-acetyl-2 3h-benzoxazolone, 2 3h-benzoxazolone, 6-acetyl, jpb 2, 6-acetyl-2-benzoxazolinone, 6-acetylbenzo d oxazol-2 3h-one, 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one, acetyl-6 benzoxazolinone french, 6-acetyl-3h-benzooxazol-2-one, 6-acetyl-1,3-benzoxazol-2 3h-one, 6-acetyl-2-oxo-3-hydrobenzoxazole | |
| CC(=O)C1=CC2=C(C=C1)NC(=O)O2 | |
| 177.159 | |
| 177.16 | |
| 6-Acetyl-2(3H)-benzoxazolone |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
RTECSNumber : DM5285500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only