missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-1-bromo-2-methoxynaphthalene 98.0+%, TCI America™

Catalog No. A15845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15845G Supplier TCI America Supplier No. A15845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84167-74-8
Molecular Formula C13H11BrO2
Molecular Weight (g/mol) 279.133
MDL Number MFCD00100832
InChI Key KDDYYAXVVDTNND-UHFFFAOYSA-N
Synonym 5′C-Bromo-6′C-methoxy-2′C-acetonaphthone
PubChem CID 622178
IUPAC Name 1-(5-bromo-6-methoxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C(=C(C=C2)OC)Br

Specifications

Quantity 5 g
Formula Weight 279.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetyl-1-bromo-2-methoxynaphthalene