accessibility menu, dialog, popup

UserName

CAS RN 84167-74-8

IUPAC Name: 1-(5-bromo-6-methoxynaphthalen-2-yl)ethan-1-one
Synonyms: 1-(5-bromo-6-methoxynaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 279.13
Molecular Formula: C13H11BrO2
InChi Key: KDDYYAXVVDTNND-UHFFFAOYSA-N
SMILES: COC1=C(Br)C2=CC=C(C=C2C=C1)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results