missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1-tetralone, 97%

Catalog No. p-4229182
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440200010 1 g
AC440200050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440200010 Supplier Thermo Scientific Chemicals Supplier No. 440200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13575-75-2 | C12H14O3 | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13575-75-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00134100
InChI Key YNNJHKOXXBIJKK-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843
PubChem CID 266816
IUPAC Name 6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1OC)C(=O)CCC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 6, 7-Dimethoxy-1-tetralone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only