accessibility menu, dialog, popup

UserName

CAS RN 13575-75-2

IUPAC Name: 6,7-dimethoxy-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: YNNJHKOXXBIJKK-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1OC)C(=O)CCC2
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)