missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol 98.0+%, TCI America™

Catalog No. B28661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28661G 1 g
B28665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28661G Supplier TCI America Supplier No. B28661G
Only null left

Chemical Identifiers

CAS 482627-84-9
Molecular Formula C24H28O3Si
Molecular Weight (g/mol) 392.57
InChI Key TUGCKICZDBMMPM-UHFFFAOYSA-N
Synonym [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane
PubChem CID 44629826
IUPAC Name [3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 392.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol