accessibility menu, dialog, popup

UserName

CAS RN 482627-84-9

CH3CH3H3CSiOOHHO

CAS RN 482627-84-9

IUPAC Name: {3-[(tert-butyldiphenylsilyl)oxy]-5-(hydroxymethyl)phenyl}methanol
Synonyms: {3-[(tert-butyldiphenylsilyl)oxy]-5-(hydroxymethyl)phenyl}methanol
Molecular Weight (g/mol): 392.57
Molecular Formula: C24H28O3Si
InChi Key: TUGCKICZDBMMPM-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](OC1=CC(CO)=CC(CO)=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results