missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-2-iodo-1,3-dimethylbenzene, 97%

Catalog No. p-7028069
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2519503 1 g
AAB2519506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2519503 Supplier Thermo Scientific Chemicals Supplier No. B2519503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5122-20-3 | C12H17I | 288.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5122-20-3
Molecular Formula C12H17I
Molecular Weight (g/mol) 288.17
MDL Number MFCD00052173
InChI Key SCKWYPTZVFBKKW-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-iodo-1,3-dimethylbenzene, maybridge4_003980, acmc-20am7p, 5-tert-butyl-2-iodo-m-xylene, 4-tert-butyl-2,6-dimethyliodobenzene, 5-tert-butyl-2-iodo-1,3-dimeth, 5-tert-butyl-2-iodo-1,3-dimethyl-benzene, benzene,5-1,1-dimethylethyl-2-iodo-1,3-dimethyl
PubChem CID 2739988
IUPAC Name 5-tert-butyl-2-iodo-1,3-dimethylbenzene
SMILES CC1=CC(=CC(C)=C1I)C(C)(C)C

Specifications

Melting Point 54°C to 57°C
Odor Odorless
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 288.17
Percent Purity 97%
Chemical Name or Material 5-tert-Butyl-2-iodo-1,3-dimethylbenzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only