CAS RN 5122-20-3
IUPAC Name:
5-tert-butyl-2-iodo-1,3-dimethylbenzene
Synonyms:
5-tert-butyl-2-iodo-1,3-dimethylbenzene
Molecular Weight (g/mol):
288.17
Molecular Formula:
C12H17I
InChi Key:
SCKWYPTZVFBKKW-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1I)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
5-tert-Butyl-2-iodo-1,3-dimethylbenzene, 97%
5-tert-Butyl-2-iodo-1,3-dimethylbenzene, 97%
CAS: 5122-20-3 Molecular Formula: C12H17I Molecular Weight (g/mol): 288.17 MDL Number: MFCD00052173 InChI Key: SCKWYPTZVFBKKW-UHFFFAOYSA-N Synonym: 5-tert-butyl-2-iodo-1,3-dimethylbenzene,maybridge4_003980,acmc-20am7p,5-tert-butyl-2-iodo-m-xylene,4-tert-butyl-2,6-dimethyliodobenzene,5-tert-butyl-2-iodo-1,3-dimeth,5-tert-butyl-2-iodo-1,3-dimethyl-benzene,benzene,5-1,1-dimethylethyl-2-iodo-1,3-dimethyl PubChem CID: 2739988 IUPAC Name: 5-tert-butyl-2-iodo-1,3-dimethylbenzene SMILES: CC1=CC(=CC(C)=C1I)C(C)(C)C