missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid, Dihydrate, Technical, 98%, Spectrum™ Chemical

Catalog No. 18608288
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608288 50 kg Amber Glass Bottle
18608285 500 g Amber Glass Bottle
18608286 2.5 kg Amber Glass Bottle
18608287 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608288 Supplier Spectrum Chemical Mfg Cor Supplier No. S171150KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5965-83-3

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 0.98
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.