missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid, Dihydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical

Catalog No. p-7166417
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608280 125 g Amber Glass Bottle
18608281 500 g Amber Glass Bottle
18608282 2.5 kg Amber Glass Bottle
18608283 12 kg Amber Glass Bottle
18608284 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608280 Supplier Spectrum Chemical Mfg Cor Supplier No. S1710125GM
Only null left

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 125 g
Percent Purity 99 to 101%
Grade ACS
Insoluble Matter 0.0002
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.