missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid, Dihydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical

Catalog No. 18608284
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608284 50 kg Amber Glass Bottle
18608280 125 g Amber Glass Bottle
18608281 500 g Amber Glass Bottle
18608282 2.5 kg Amber Glass Bottle
18608283 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608284 Supplier Spectrum Chemical Mfg Cor Supplier No. S171050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 99 to 101%
Grade ACS
Insoluble Matter 0.0002
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.